1-(2,6-Dichloropyridin-3-yl)-2,2,2-trifluoroethanol ≥97%

Supplier: AOBChem USA

Warning

36203-500MG 36203-1G 36203-5G
77336-780EA 394.49 USD
77336-780 77349-766 77373-968
1-(2,6-Dichloropyridin-3-yl)-2,2,2-trifluoroethanol ≥97%
1-(2,6-Dichloropyridin-3-yl)-2,2,2-trifluoroethanol

Precaución: for Research Use Only

Formula: C₇H₄Cl₂F₃NO
MW: 246.014 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28059702
CAS Number: 1823358-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR