6-Chloro-3-fluoro-2-(oxazol-5-yl)phenol ≥95%
Supplier: AOBChem USA
|
Warning
|
36187-500MG
36187-1G
36187-5G
77339-350EA
581.95
USD
77339-350
77349-762
77373-966
6-Chloro-3-fluoro-2-(oxazol-5-yl)phenol ≥95%
6-Chloro-3-fluoro-2-(oxazol-5-yl)phenol
Precaución: for Research Use Only
Formula:
C₉H₅ClFNO₂ MW: 213.593 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32631978 CAS Number: 2379321-38-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...