4-Methyl-2-(oxazol-5-yl)phenol ≥95%

Supplier: AOBChem USA

Warning

36186-500MG 36186-1G 36186-5G
77335-790EA 366.68 USD
77335-790 77349-760 77373-964
4-Methyl-2-(oxazol-5-yl)phenol ≥95%
4-Methyl-2-(oxazol-5-yl)phenol

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD32631977
CAS Number: 2379322-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR