Ethyl 2-bromo-6-fluoro-4-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

36182-500MG 36182-1G 36182-5G 36182-25G
77326-068EA 218.36 USD
77326-068 77349-758 77362-382 77385-330
Ethyl 2-bromo-6-fluoro-4-methylbenzoate ≥97%
Ethyl 2-bromo-6-fluoro-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrFO₂
MW: 261.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28503345
CAS Number: 1807088-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR