3-Benzyloxy-2-fluorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(3-(Benzyloxy)-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Benzyloxy-2-fluorobenzeneboronic acid pinacol ester

36179-500MG 36179-1G 36179-5G 36179-25G
77326-066EA 188.49 USD
77326-066 77349-756 77362-380 77385-328
3-Benzyloxy-2-fluorophenylboronic acid pinacol ester ≥97%
3-Benzyloxy-2-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₉H₂₂BFO₃
Storage Temperature: Ambient
MDL Number: MFCD22419264
CAS Number: 874152-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR