tert-Butyl 3-bromo-4-chloro-5-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

36177-500MG 36177-1G 36177-5G
77337-486EA 415.09 USD
77337-486 77349-754 77373-962
tert-Butyl 3-bromo-4-chloro-5-fluorobenzoate ≥97%
tert-Butyl 3-bromo-4-chloro-5-fluorobenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrClFO₂
Storage Temperature: Ambient
MDL Number: MFCD32631974
CAS Number: 2379321-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR