(4-(Trifluoromethoxy)-[1,1'-biphenyl]-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

36173-500MG 36173-1G 36173-5G
77338-328EA 477.92 USD
77338-328 77349-750 77373-958
(4-(Trifluoromethoxy)-[1,1'-biphenyl]-2-yl)methanol ≥95%
(4-(Trifluoromethoxy)-[1,1'-biphenyl]-2-yl)methanol

Precaución: for Research Use Only

Formula: C₁₄H₁₁F₃O₂
MW: 268.231 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631970
CAS Number: 2379321-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR