4-Bromo-2,5-difluoro-N-methoxy-N-methylbenzamide ≥95%

Supplier: AOBChem USA

Warning

36172-500MG 36172-1G 36172-5G 36172-25G
77326-064EA 144.2 USD
77326-064 77345-046 77362-378 77385-326
4-Bromo-2,5-difluoro-N-methoxy-N-methylbenzamide ≥95%
4-Bromo-2,5-difluoro-N-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₈BrF₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD32631969
CAS Number: 2007959-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR