4-Bromo-N-cyclohexyl-2,5-difluorobenzamide ≥97%

Supplier: AOBChem USA

Warning

36170-500MG 36170-1G 36170-5G 36170-25G
77326-062EA 243.08 USD
77326-062 77349-748 77362-376 77385-324
4-Bromo-N-cyclohexyl-2,5-difluorobenzamide ≥97%
4-Bromo-N-cyclohexyl-2,5-difluorobenzamide

Precaución: for Research Use Only

Formula: C₁₃H₁₄BrF₂NO
MW: 318.157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631967
CAS Number: 2379322-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR