4-Bromo-N-cyclopropyl-2,5-difluorobenzamide ≥97%

Supplier: AOBChem USA

Warning

36169-500MG 36169-1G 36169-5G 36169-25G
77326-060EA 243.08 USD
77326-060 77349-746 77362-374 77385-322
4-Bromo-N-cyclopropyl-2,5-difluorobenzamide ≥97%
4-Bromo-N-cyclopropyl-2,5-difluorobenzamide

Precaución: for Research Use Only

Formula: C₁₀H₈BrF₂NO
MW: 276.077 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631966
CAS Number: 2379322-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR