2-Ethoxy-3-fluorophenol ≥95%

Supplier: AOBChem USA

Warning

36158-500MG 36158-1G 36158-5G 36158-25G
77326-056EA 247.2 USD
77326-056 77349-742 77362-370 77385-318
2-Ethoxy-3-fluorophenol ≥95%
2-Ethoxy-3-fluorophenol

Precaución: for Research Use Only

Formula: C₈H₉FO₂
MW: 156.154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23111044
CAS Number: 909302-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR