5-(4-Bromo-2-chlorophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

36120-500MG 36120-1G 36120-5G
77336-348EA 387.28 USD
77336-348 77349-738 77373-956
5-(4-Bromo-2-chlorophenyl)oxazole ≥97%
5-(4-Bromo-2-chlorophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₅BrClNO
MW: 258.499 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31556575
CAS Number: 2002638-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR