4-Bromo-1-(4-chloro-2-fluorophenyl)-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

36108-500MG 36108-1G 36108-5G 36108-25G
77326-050EA 237.93 USD
77326-050 77349-736 77362-364 77385-312
4-Bromo-1-(4-chloro-2-fluorophenyl)-1H-pyrazole ≥97%
4-Bromo-1-(4-chloro-2-fluorophenyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₉H₅BrClFN₂
MW: 275.505 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631951
CAS Number: 2379322-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR