1-(4-Chloro-2-fluorophenyl)-1H-pyrazole-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

36106-500MG 36106-1G 36106-5G
77337-772EA 441.87 USD
77337-772 77349-734 77373-954
1-(4-Chloro-2-fluorophenyl)-1H-pyrazole-4-carbaldehyde ≥97%
1-(4-Chloro-2-fluorophenyl)-1H-pyrazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₆ClFN₂O
MW: 224.619 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631949
CAS Number: 2379321-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR