1-(4-Bromo-3-methylphenyl)-3,5-dimethyl-1H-pyrazole-4-carbaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
36099-500MG
36099-1G
36099-5G
36099-25G
77326-046EA
233.81
USD
77326-046
77349-732
77362-360
77385-310
1-(4-Bromo-3-methylphenyl)-3,5-dimethyl-1H-pyrazole-4-carbaldehyde ≥97%
1-(4-Bromo-3-methylphenyl)-3,5-dimethyl-1H-pyrazole-4-carbaldehyde
Precaución: for Research Use Only
Formula:
C₁₃H₁₃BrN₂O MW: 293.159 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11959837 CAS Number: 1176631-90-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...