5-Chloro-N-cyclopropylpicolinamide ≥97%

Supplier: AOBChem USA

Warning

36090-500MG 36090-1G 36090-5G
77326-042EA 348.14 USD
77326-042 77349-728 77373-948
5-Chloro-N-cyclopropylpicolinamide ≥97%
5-Chloro-N-cyclopropylpicolinamide

Precaución: for Research Use Only

Formula: C₉H₉ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD20361639
CAS Number: 1289212-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR