N-Cyclopropyl-2,3-difluorobenzamide ≥97%

Supplier: AOBChem USA

Warning

36042-1G 36042-5G 36042-25G
77341-914EA 108.15 USD
77341-914 77362-342 77373-936
N-Cyclopropyl-2,3-difluorobenzamide ≥97%
N-Cyclopropyl-2,3-difluorobenzamide

Precaución: for Research Use Only

Formula: C₁₀H₉F₂NO
MW: 197.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16836079
CAS Number: 1247630-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR