(2,3-difluorophenyl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

36038-1G 36038-5G 36038-25G
77341-476EA 89.61 USD
77341-476 77362-334 77373-928
(2,3-difluorophenyl)(thiomorpholino)methanone ≥97%
(2,3-difluorophenyl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₁F₂NOS
MW: 243.273 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16836151
CAS Number: 1249010-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR