(2,3-difluorophenyl)(morpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

36037-1G 36037-5G 36037-25G
77341-672EA 99.91 USD
77341-672 77362-332 77373-926
(2,3-difluorophenyl)(morpholino)methanone ≥97%
(2,3-difluorophenyl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₁F₂NO₂
MW: 227.207 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00543905
CAS Number: 351038-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR