2,3-Difluoro-N,N-dimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

36036-500MG 36036-1G 36036-5G 36036-25G
77326-030EA 187.46 USD
77326-030 77349-722 77362-330 77385-298
2,3-Difluoro-N,N-dimethylbenzamide ≥97%
2,3-Difluoro-N,N-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₉F₂NO
Storage Temperature: Ambient
MDL Number: MFCD16836100
CAS Number: 1250280-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR