Isopropyl 2-chloro-5-fluoro-3-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

36033-500MG 36033-1G 36033-5G
77336-598EA 389.34 USD
77336-598 77349-720 77373-924
Isopropyl 2-chloro-5-fluoro-3-(trifluoromethyl)benzoate ≥97%
Isopropyl 2-chloro-5-fluoro-3-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₁H₉ClF₄O₂
MW: 284.635 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631938
CAS Number: 2364585-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR