Methyl 4-chloro-3-(difluoromethoxy)benzoate ≥97%

Supplier: AOBChem USA

Warning

36023-500MG 36023-1G 36023-5G
77338-312EA 475.86 USD
77338-312 77349-716 77373-920
Methyl 4-chloro-3-(difluoromethoxy)benzoate ≥97%
Methyl 4-chloro-3-(difluoromethoxy)benzoate

Precaución: for Research Use Only

Formula: C₉H₇ClF₂O₃
MW: 236.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28750643
CAS Number: 1807030-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR