1-Bromo-3-(difluoromethoxy)-5-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

36018-500MG 36018-1G 36018-5G 36018-25G
77326-028EA 132.87 USD
77326-028 77344-400 77362-328 77385-296
1-Bromo-3-(difluoromethoxy)-5-methoxybenzene ≥97%
1-Bromo-3-(difluoromethoxy)-5-methoxybenzene

Precaución: for Research Use Only

Formula: C₈H₇BrF₂O₂
MW: 253.041 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18390869
CAS Number: 1261446-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR