5-(2,6-Dichloro-4-fluorophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

36016-500MG 36016-1G 36016-5G
77336-596EA 389.34 USD
77336-596 77349-714 77373-918
5-(2,6-Dichloro-4-fluorophenyl)oxazole ≥97%
5-(2,6-Dichloro-4-fluorophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₄Cl₂FNO
MW: 232.039 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631933
CAS Number: 2364585-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR