5-(3,5-Dibromophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

36014-500MG 36014-1G 36014-5G 36014-25G
77326-026EA 129.78 USD
77326-026 77344-308 77362-326 77373-916
5-(3,5-Dibromophenyl)oxazole ≥97%
5-(3,5-Dibromophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₅Br₂NO
MW: 302.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631932
CAS Number: 2024443-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR