5-(3,5-Dibromo-4-methylphenyl)oxazole ≥95%

Supplier: AOBChem USA

Warning

36012-500MG 36012-1G 36012-5G
77326-024EA 311.06 USD
77326-024 77349-712 77373-914
5-(3,5-Dibromo-4-methylphenyl)oxazole ≥95%
5-(3,5-Dibromo-4-methylphenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₀H₇Br₂NO
MW: 316.977 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631930
CAS Number: 2364584-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR