5-(4-Bromo-3-methoxyphenyl)oxazole ≥95%

Supplier: AOBChem USA

Warning

36009-500MG 36009-1G 36009-5G 36009-25G
77326-020EA 259.56 USD
77326-020 77349-710 77362-322 77385-292
5-(4-Bromo-3-methoxyphenyl)oxazole ≥95%
5-(4-Bromo-3-methoxyphenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD32631927
CAS Number: 2379321-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR