5-(5-Bromo-2-methoxyphenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

36008-500MG 36008-1G 36008-5G 36008-25G
77326-018EA 150.38 USD
77326-018 77345-384 77362-320 77385-290
5-(5-Bromo-2-methoxyphenyl)oxazole ≥97%
5-(5-Bromo-2-methoxyphenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO₂
MW: 254.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28125782
CAS Number: 1091618-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR