5-(4-Chloro-3-(trifluoromethyl)phenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

36001-500MG 36001-1G 36001-5G 36001-25G
77326-014EA 134.93 USD
77326-014 77344-460 77362-318 77385-288
5-(4-Chloro-3-(trifluoromethyl)phenyl)oxazole ≥97%
5-(4-Chloro-3-(trifluoromethyl)phenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₀H₅ClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD32631920
CAS Number: 2379322-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR