5-(4-Chloro-2-(trifluoromethyl)phenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35999-500MG 35999-1G 35999-5G 35999-25G
77326-012EA 156.56 USD
77326-012 77345-624 77362-316 77385-286
5-(4-Chloro-2-(trifluoromethyl)phenyl)oxazole ≥97%
5-(4-Chloro-2-(trifluoromethyl)phenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₀H₅ClF₃NO
MW: 247.601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631918
CAS Number: 2142915-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR