5-(4-Bromo-2,5-difluorophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35998-500MG 35998-1G 35998-5G 35998-25G
77326-010EA 261.62 USD
77326-010 77349-704 77362-314 77385-284
5-(4-Bromo-2,5-difluorophenyl)oxazole ≥97%
5-(4-Bromo-2,5-difluorophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₄BrF₂NO
Storage Temperature: Ambient
MDL Number: MFCD32631917
CAS Number: 2364584-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR