5-(2-Fluoro-3-methoxyphenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35997-500MG 35997-1G 35997-5G 35997-25G
77326-008EA 156.56 USD
77326-008 77345-622 77362-312 77385-282
5-(2-Fluoro-3-methoxyphenyl)oxazole ≥97%
5-(2-Fluoro-3-methoxyphenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₀H₈FNO₂
Storage Temperature: Ambient
MDL Number: MFCD32631916
CAS Number: 1996833-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR