5-(4-(benzyloxy)-2,3-difluorophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35996-500MG 35996-1G 35996-5G
77339-400EA 586.07 USD
77339-400 77349-702 77373-908
5-(4-(benzyloxy)-2,3-difluorophenyl)oxazole ≥97%
5-(4-(benzyloxy)-2,3-difluorophenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₆H₁₁F₂NO₂
MW: 287.261 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631915
CAS Number: 2364584-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR