5-(2-Bromo-3,6-dichlorophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35995-500MG 35995-1G 35995-5G
77337-484EA 415.09 USD
77337-484 77349-700 77373-906
5-(2-Bromo-3,6-dichlorophenyl)oxazole ≥97%
5-(2-Bromo-3,6-dichlorophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₄BrCl₂NO
MW: 292.944 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631914
CAS Number: 2364585-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR