5-(2,5-Dibromophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35994-500MG 35994-1G 35994-5G 35994-25G
77326-006EA 182.31 USD
77326-006 77346-938 77362-310 77385-280
5-(2,5-Dibromophenyl)oxazole ≥97%
5-(2,5-Dibromophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₅Br₂NO
MW: 302.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631913
CAS Number: 2013263-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR