5-(2-Methoxy-5-(trifluoromethyl)phenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35992-500MG 35992-1G 35992-5G 35992-25G
77326-004EA 156.56 USD
77326-004 77345-620 77362-308 77385-278
5-(2-Methoxy-5-(trifluoromethyl)phenyl)oxazole ≥97%
5-(2-Methoxy-5-(trifluoromethyl)phenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₁H₈F₃NO₂
MW: 243.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631911
CAS Number: 2364584-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR