5-(3-Bromo-5-methoxyphenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35990-500MG 35990-1G 35990-5G 35990-25G
77326-002EA 208.06 USD
77326-002 77349-698 77362-306 77385-276
5-(3-Bromo-5-methoxyphenyl)oxazole ≥97%
5-(3-Bromo-5-methoxyphenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD32631909
CAS Number: 2364584-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR