5-(3-(benzyloxy)-4-fluorophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35989-500MG 35989-1G 35989-5G
77339-568EA 597.4 USD
77339-568 77349-696 77373-904
5-(3-(benzyloxy)-4-fluorophenyl)oxazole ≥97%
5-(3-(benzyloxy)-4-fluorophenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₆H₁₂FNO₂
MW: 269.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631908
CAS Number: 2364585-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR