5-(4-Bromo-3,5-dimethylphenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35985-500MG 35985-1G 35985-5G
77336-594EA 389.34 USD
77336-594 77349-692 77373-900
5-(4-Bromo-3,5-dimethylphenyl)oxazole ≥97%
5-(4-Bromo-3,5-dimethylphenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrNO
MW: 252.107 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631904
CAS Number: 2225759-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR