5-(4-Bromo-3-fluorophenyl)oxazole ≥95%

Supplier: AOBChem USA

Warning

35983-500MG 35983-1G 35983-5G 35983-25G
77326-000EA 129.78 USD
77326-000 77344-306 77362-304 77373-898
5-(4-Bromo-3-fluorophenyl)oxazole ≥95%
5-(4-Bromo-3-fluorophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₅BrFNO
MW: 242.045 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631902
CAS Number: 2111410-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR