5-(2,3-Dichloro-6-(trifluoromethyl)phenyl)oxazole ≥95%

Supplier: AOBChem USA

Warning

35981-500MG 35981-1G 35981-5G 35981-25G
77325-996EA 238.96 USD
77325-996 77349-688 77362-300 77385-272
5-(2,3-Dichloro-6-(trifluoromethyl)phenyl)oxazole ≥95%
5-(2,3-Dichloro-6-(trifluoromethyl)phenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₀H₄Cl₂F₃NO
MW: 282.046 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631900
CAS Number: 2364584-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR