5-(2-Chloro-4-(trifluoromethyl)phenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35980-500MG 35980-1G 35980-5G
77325-994EA 311.06 USD
77325-994 77349-686 77373-896
5-(2-Chloro-4-(trifluoromethyl)phenyl)oxazole ≥97%
5-(2-Chloro-4-(trifluoromethyl)phenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₀H₅ClF₃NO
MW: 247.601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631899
CAS Number: 2364584-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR