5-(5-Chloro-2-(trifluoromethyl)phenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35979-500MG 35979-1G 35979-5G 35979-25G
77325-992EA 182.31 USD
77325-992 77346-936 77362-298 77385-270
5-(5-Chloro-2-(trifluoromethyl)phenyl)oxazole ≥97%
5-(5-Chloro-2-(trifluoromethyl)phenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₀H₅ClF₃NO
MW: 247.601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631898
CAS Number: 2364584-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR