5-(2-Bromo-3-chloro-6-fluorophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35978-500MG 35978-1G 35978-5G
77338-562EA 498.52 USD
77338-562 77349-684 77373-894
5-(2-Bromo-3-chloro-6-fluorophenyl)oxazole ≥97%
5-(2-Bromo-3-chloro-6-fluorophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₄BrClFNO
MW: 276.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631897
CAS Number: 2364585-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR