5-(3-Bromo-5-chlorophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35977-500MG 35977-1G 35977-5G 35977-25G
77325-990EA 238.96 USD
77325-990 77349-682 77362-296 77385-268
5-(3-Bromo-5-chlorophenyl)oxazole ≥97%
5-(3-Bromo-5-chlorophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₅BrClNO
MW: 258.499 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631896
CAS Number: 2011450-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR