5-(3-Bromo-2-fluorophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35976-500MG 35976-1G 35976-5G 35976-25G
77325-988EA 182.31 USD
77325-988 77346-934 77362-294 77385-266
5-(3-Bromo-2-fluorophenyl)oxazole ≥97%
5-(3-Bromo-2-fluorophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₅BrFNO
MW: 242.045 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631895
CAS Number: 2017304-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR