5-(2-Bromo-6-fluoro-3-methoxyphenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35972-500MG 35972-1G 35972-5G
77336-592EA 389.34 USD
77336-592 77349-680 77373-892
5-(2-Bromo-6-fluoro-3-methoxyphenyl)oxazole ≥97%
5-(2-Bromo-6-fluoro-3-methoxyphenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₀H₇BrFNO₂
MW: 272.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631891
CAS Number: 2364585-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR