5-(2,4-Dibromo-6-fluorophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35971-500MG 35971-1G 35971-5G
77338-560EA 498.52 USD
77338-560 77349-678 77373-890
5-(2,4-Dibromo-6-fluorophenyl)oxazole ≥97%
5-(2,4-Dibromo-6-fluorophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₄Br₂FNO
MW: 320.941 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631890
CAS Number: 2364585-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR