5-(2-Bromo-3,6-difluorophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35970-500MG 35970-1G 35970-5G 35970-25G
77325-984EA 259.56 USD
77325-984 77349-676 77362-290 77385-262
5-(2-Bromo-3,6-difluorophenyl)oxazole ≥97%
5-(2-Bromo-3,6-difluorophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₄BrF₂NO
MW: 260.035 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631889
CAS Number: 2364585-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR