5-(2-Fluoro-5-(trifluoromethyl)phenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35968-500MG 35968-1G 35968-5G 35968-25G
77325-982EA 134.93 USD
77325-982 77344-458 77362-288 77385-260
5-(2-Fluoro-5-(trifluoromethyl)phenyl)oxazole ≥97%
5-(2-Fluoro-5-(trifluoromethyl)phenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₀H₅F₄NO
MW: 231.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631887
CAS Number: 2026425-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR